Gene Transcripts

Gene Symbol
KRTAP5-9
Entrez Gene ID
3846
Gene Name
keratin associated protein 5-9
Gene Aliases
KRN1, KRTAP5-1, KRTAP5.9
Chromosome Location
Chr. 11: 71548420 - 71549607 on Build GRCh38
UniGene ID
Hs.709220
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005553.3 NP_005544.4 1 909
GenBank mRNA AB126078.1 - 1 705
X63755.1 - 1 909

View Full Product Details