Gene Transcripts

Gene Symbol
Asah2
Entrez Gene ID
54447
Gene Name
N-acylsphingosine amidohydrolase 2
Gene Aliases
AI585898
Chromosome Location
Chr. 19: 31982597 - 32108233 on Build GRCm38
UniGene ID
Mm.491229
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018830.1 NP_061300.1 23 2494
GenBank mRNA AB037111.1 - 24 2779
AB037181.1 - 23 2494
AK080951.1 - 23 2671
AK136189.1 - 1 44
BC022604.1 - 14 1276
BC137898.1 - 23 2477 8860917
BC145016.1 - 23 2477 9053564

View Full Product Details