Gene Transcripts

Gene Symbol
Apbb1ip
Entrez Gene ID
54519
Gene Name
amyloid beta (A4) precursor protein-binding, family B, member 1 interacting protein
Gene Aliases
9930118P07Rik, Prp48
Chromosome Location
Chr. 2: 22755989 - 22875653 on Build GRCm38
UniGene ID
Mm.14255
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019456.2 NP_062329.2 12, 13 1579
GenBank mRNA AF020313.1 - 9, 10 1184
AK089987.1 - 4, 5 453
BC023110.1 - 12, 13 1468 3707478

View Full Product Details