Gene Transcripts
- Gene Symbol
- Trim3
- Entrez Gene ID
- 55992
- Gene Name
- tripartite motif-containing 3
- Gene Aliases
- BERP1, HAC1, Rnf22
- Chromosome Location
- Chr. 7: 105604463 - 105633571 on Build GRCm38
- UniGene ID
- Mm.29290
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001285870.1 |
NP_001272799.1
|
7, 8 |
1756 |
|
NM_001285871.1 |
NP_001272800.1
|
7, 8 |
1941 |
|
NM_001285873.1 |
NP_001272802.1
|
7, 8 |
1756 |
|
NM_018880.3 |
NP_061368.1
|
7, 8 |
1730 |
|
GenBank mRNA |
AB030912.1 |
- |
7, 8 |
1757 |
|
AF220019.1 |
- |
7, 8 |
1649 |
|
AK019165.1 |
- |
7, 8 |
1691 |
|
AK144674.1 |
- |
7, 8 |
1659 |
|
AK170029.1 |
- |
7, 8 |
1694 |
|
BC003347.1 |
- |
7, 8 |
1770 |
|
BC034263.1 |
- |
7, 8 |
1695 |
4503884 |
View Full Product Details