Gene Transcripts

Gene Symbol
Nagk
Entrez Gene ID
56174
Gene Name
N-acetylglucosamine kinase
Gene Aliases
Gnk
Chromosome Location
Chr. 6: 83795031 - 83802556 on Build GRCm38
UniGene ID
Mm.190650
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164187.1 NP_001157659.1 5 578
NM_019542.2 NP_062415.1 5 587
GenBank mRNA AJ242909.1 - 5 599
AJ242911.1 - 5 539
AK007240.1 - 5 582
AK155292.1 - 5 479
BC004689.1 - 5 465 3501470

View Full Product Details