Gene Transcripts

Gene Symbol
Msgn1
Entrez Gene ID
56184
Gene Name
mesogenin 1
Gene Aliases
Msgn, pMsgn1
Chromosome Location
Chr. 12: 11208382 - 11208948 on Build GRCm38
UniGene ID
Mm.117113
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019544.1 NP_062417.1 1 486
GenBank mRNA AF261105.1 - 1 486
BC116996.1 - 1 510 40126315

View Full Product Details