Gene Transcripts

Gene Symbol
Dhodh
Entrez Gene ID
56749
Gene Name
dihydroorotate dehydrogenase
Gene Aliases
2810417D19Rik, AI834883
Chromosome Location
Chr. 8: 109593244 - 109608701 on Build GRCm38
UniGene ID
Mm.23894
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020046.3 NP_064430.1 3 616
GenBank mRNA AF029667.2 - 3 321
AK013102.1 - 3 568
AK030798.1 - 106
AK088863.1 - 3 338
AK149350.1 - 3 364
BC019542.1 - 3 326 4039461
BC027829.1 - 3 577 5253723
BC045206.1 - 3 566 5253764

View Full Product Details