Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020560.2 | NP_065585.1 | 2 | 289 | |
GenBank mRNA | AK005110.1 | - | 2 | 248 | |
AK011825.1 | - | 2 | 245 | ||
AK019202.1 | - | 2 | 269 | ||
AK078073.1 | - | 2 | 232 | ||
AK161913.1 | - | 2 | 232 | ||
BC006764.1 | - | 2 | 184 | ||
BC018475.1 | - | 2 | 188 | 3492633 | |
BC027430.1 | - | 2 | 193 | 4187438 | |
BY237441.1 | - | 2 | 291 | ||
Z46966.1 | - | 2 | 208 |