Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021524.2 | NP_067499.2 | 8 | 1307 | |
GenBank mRNA | AF234625.1 | - | 8 | 1167 | |
AK086415.1 | - | 8 | 1138 | ||
AK153926.1 | - | 8 | 1138 | ||
AK166270.1 | - | 8 | 1167 | ||
AY679720.1 | - | 8 | 1142 | ||
BC004059.1 | - | 8 | 1115 | 3590087 | |
BC018358.1 | - | 8 | 1307 | 2609271 | |
CV561474.1 | - | 2 | 164 |