Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025511.2 | NP_079787.1 | 3, 4 | 236 | |
GenBank mRNA | AK009178.1 | - | 3, 4 | 230 | |
AK019293.1 | - | 2, 3 | 128 | ||
BC055277.1 | - | 3, 4 | 219 | 5714061 | |
BC059717.1 | - | 3, 4 | 234 | 6543216 | |
BY779933.1 | - | 3, 4 | 236 |