Gene Transcripts
- Gene Symbol
- Diablo
- Entrez Gene ID
- 66593
- Gene Name
- diablo homolog (Drosophila)
- Gene Aliases
- 0610041G12Rik, 1700006L01Rik, AU040403, Smac
- Chromosome Location
- Chr. 5: 123511330 - 123524164 on Build GRCm38
- UniGene ID
- Mm.46716
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023232.3 |
NP_075721.3
|
2, 3 |
213 |
|
GenBank mRNA |
AF203914.1 |
- |
2, 3 |
179 |
|
AK002887.1 |
- |
2, 3 |
183 |
|
AK012760.1 |
- |
2, 3 |
211 |
|
AK077198.1 |
- |
2, 3 |
194 |
|
AK082903.1 |
- |
2, 3 |
183 |
|
AK133194.1 |
- |
|
193 |
|
AK169573.1 |
- |
2, 3 |
183 |
|
BC024780.1 |
- |
2, 3 |
185 |
5365266 |
BC054109.1 |
- |
2, 3 |
207 |
30013833 |
BY080746.1 |
- |
2, 3 |
225 |
|
View Full Product Details