Gene Transcripts

Gene Symbol
Cntnap2
Entrez Gene ID
66797
Gene Name
contactin associated protein-like 2
Gene Aliases
5430425M22Rik, Caspr2, mKIAA0868
Chromosome Location
Chr. 6: 45060061 - 47301388 on Build GRCm38
UniGene ID
Mm.440084
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004357.2 NP_001004357.2 24 4365
NM_025771.3 NP_080047.1 4 812
GenBank mRNA AK013139.1 - 4 780
AK017341.1 - 4 807
AK129232.1 - 24 4338
BC029826.1 - 4 761
BC064467.1 - 19 3256
BC103799.1 - 4 719

View Full Product Details