Gene Transcripts

Gene Symbol
Rbm22
Entrez Gene ID
66810
Gene Name
RNA binding motif protein 22
Gene Aliases
8430430L24Rik
Chromosome Location
Chr. 18: 60560786 - 60572729 on Build GRCm38
UniGene ID
Mm.275106
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025776.2 NP_080052.1 9 1013
GenBank mRNA AK018452.1 - 9 1036
AK045116.1 - 9 1033
AK150271.1 - 9 1026
AK151758.1 - 9 1026
AK152252.1 - 9 1026
AK166600.1 - 9 1034
AK167509.1 - 9 1053
AK168919.1 - 9 1014
BC042691.1 - 9 1032
BC080205.1 - 9 1011 3709760

View Full Product Details