Gene Transcripts

Gene Symbol
Ndufb7
Entrez Gene ID
66916
Gene Name
NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 7
Gene Aliases
1110002H15Rik
Chromosome Location
Chr. 8: 83566758 - 83571623 on Build GRCm38
UniGene ID
Mm.29683
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025843.3 NP_080119.1 3 365
GenBank mRNA AK002678.1 - 3 363
AK003300.1 - 3 366
BC034787.1 - 3 344 4913656
BC056984.1 - 2 4206

View Full Product Details