Gene Transcripts
- Gene Symbol
- Uqcrc2
- Entrez Gene ID
- 67003
- Gene Name
- ubiquinol cytochrome c reductase core protein 2
- Gene Aliases
- 1500004O06Rik
- Chromosome Location
- Chr. 7: 120635189 - 120659523 on Build GRCm38
- UniGene ID
- Mm.334206
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025899.2 |
NP_080175.1
|
2, 3 |
217 |
|
GenBank mRNA |
AK003975.1 |
- |
2, 3 |
190 |
|
AK004492.1 |
- |
2, 3 |
192 |
|
AK005151.1 |
- |
2, 3 |
201 |
|
AK075856.1 |
- |
2, 3 |
161 |
|
AK077583.1 |
- |
2, 3 |
166 |
|
AK088103.1 |
- |
2, 3 |
171 |
|
AK167377.1 |
- |
2, 3 |
171 |
|
BC003423.1 |
- |
2, 3 |
189 |
2609295 |
CJ074691.1 |
- |
2, 3 |
218 |
|
View Full Product Details