Gene Transcripts

Gene Symbol
Gp2
Entrez Gene ID
67133
Gene Name
glycoprotein 2 (zymogen granule membrane)
Gene Aliases
2310037I18Rik, AV060639
Chromosome Location
Chr. 7: 119442542 - 119459272 on Build GRCm38
UniGene ID
Mm.46403
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025989.3 NP_080265.2 4 699
GenBank mRNA AK009323.1 - 4 700
AK009661.1 - 4 700
AK009843.1 - 4 653
BC119573.2 - 4 694 40057995

View Full Product Details