Gene Transcripts

Gene Symbol
Wdr48
Entrez Gene ID
67561
Gene Name
WD repeat domain 48
Gene Aliases
8430408H12Rik, Uaf1, mKIAA1449
Chromosome Location
Chr. 9: 119894895 - 119926586 on Build GRCm38
UniGene ID
Mm.104789
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026236.3 NP_080512.1 2 168
GenBank mRNA AK012599.1 - 2 168
AK018396.1 - 2 171
AK030041.1 - 2 169
AK031244.1 - 2 180
AK033631.1 - 2 167
AK035273.1 - 2 170
AK035864.1 - 2 170
AK043970.1 - 2 170
AK122511.1 - 2 152
AK128953.1 - 2 168
BC006679.1 - 1 31
BC048155.1 - 2 157 5698339
BC050106.1 - 2 185
BC062967.1 - 2 172 6835444

View Full Product Details