Gene Transcripts

Gene Symbol
Mgat4c
Entrez Gene ID
67569
Gene Name
MGAT4 family, member C
Gene Aliases
9130411I17Rik, GntIVh
Chromosome Location
Chr. 10: 101681487 - 102391851 on Build GRCm38
UniGene ID
Mm.25252
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162368.1 NP_001155840.1 6 1527
NM_001162369.1 NP_001155841.1 6 1400
NM_001205098.1 NP_001192027.1 8 1940
NM_026243.4 NP_080519.2 4 1422
GenBank mRNA AK018574.1 - 4 1421
AK018672.1 - 4 1425
AK020253.1 - 1 15
AK032009.1 - 6 1403
AK033482.1 - 4 1423
BC046987.1 - 4 1410 6475851

View Full Product Details