Gene Transcripts

Gene Symbol
Eqtn
Entrez Gene ID
67753
Gene Name
equatorin, sperm acrosome associated
Gene Aliases
1700028B15Rik, 4930579C15Rik, Afaf, C81058, Eqtl, Eqts
Chromosome Location
Chr. 4: 94875696 - 94929950 on Build GRCm38
UniGene ID
Mm.25097
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290623.1 NP_001277552.1 6, 7 586
NM_027089.4 NP_081365.2 6, 7 586
GenBank mRNA AB438105.1 - 6, 7 495
AB438106.1 - 6, 7 495
AK006446.1 - 6, 7 537
AK016738.1 - 6, 7 557
BC089517.1 - 6, 7 553 6740450
BE632290.1 - 2, 3 105
BY716419.1 - 6, 7 553
DQ336137.1 - 6, 7 537
EU828781.1 - 6, 7 571
EU828782.1 - 6, 7 571

View Full Product Details