Gene Transcripts

Gene Symbol
Mum1
Entrez Gene ID
68114
Gene Name
melanoma associated antigen (mutated) 1
Gene Aliases
2610019J09, 9430059D04Rik, UBE-1C2
Chromosome Location
Chr. 10: 80226434 - 80245144 on Build GRCm38
UniGene ID
Mm.273418
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023431.5 NP_075920.4 3, 4 319
GenBank mRNA AK154610.1 - 3, 4 238
AK158750.1 - 3, 4 319
AK170459.1 - 3, 4 238
AK205449.1 - 2, 3 180
AK220121.1 - 2, 3 180
BC075617.1 - 2, 3 208 5718202

View Full Product Details