Gene Transcripts

Gene Symbol
Dhrs7c
Entrez Gene ID
68460
Gene Name
dehydrogenase/reductase (SDR family) member 7C
Gene Aliases
1110001P11Rik, AI120487, SRP-35
Chromosome Location
Chr. 11: 67798271 - 67816002 on Build GRCm38
UniGene ID
Mm.22167
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013013.2 NP_001013031.2 4 605
GenBank mRNA AI391290.1 - 1 94
BC039565.1 - 4 496
CJ048402.1 - 4 601

View Full Product Details