Gene Transcripts
- Gene Symbol
- Haghl
- Entrez Gene ID
- 68977
- Gene Name
- hydroxyacylglutathione hydrolase-like
- Gene Aliases
- 1500017E18Rik, 2810014I23Rik, C330022E15Rik
- Chromosome Location
- Chr. 17: 25779843 - 25785669 on Build GRCm38
- UniGene ID
- Mm.29230
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271433.1 |
NP_001258362.1
|
3, 4 |
413 |
|
NM_001271434.1 |
NP_001258363.1
|
3, 4 |
413 |
|
NM_001271435.1 |
NP_001258364.1
|
3, 4 |
413 |
|
NM_001271436.1 |
NP_001258365.1
|
2, 3 |
227 |
|
NM_001271437.1 |
NP_001258366.1
|
2, 3 |
222 |
|
NM_026897.3 |
NP_081173.1
|
2, 3 |
398 |
|
GenBank mRNA |
AK005274.1 |
- |
2, 3 |
394 |
|
AK012748.1 |
- |
3, 4 |
410 |
|
AK021220.1 |
- |
2, 3 |
224 |
|
AK032999.1 |
- |
2, 3 |
204 |
|
AK034798.1 |
- |
3, 4 |
388 |
|
AK047517.1 |
- |
3, 4 |
368 |
|
AK079632.1 |
- |
2, 3 |
396 |
|
AK154248.1 |
- |
2, 3 |
204 |
|
BC005586.1 |
- |
3, 4 |
359 |
|
BC030466.1 |
- |
3, 4 |
398 |
5253213 |
BC083322.1 |
- |
3, 4 |
352 |
6820885 |
BY265614.1 |
- |
3, 4 |
414 |
|
View Full Product Details