Gene Transcripts

Gene Symbol
Anapc13
Entrez Gene ID
69010
Gene Name
anaphase promoting complex subunit 13
Gene Aliases
1810004D07Rik, APC13, SWM1
Chromosome Location
Chr. 9: 102626296 - 102634244 on Build GRCm38
UniGene ID
Mm.2000
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181394.3 NP_852059.1 3 209
GenBank mRNA AK007332.1 - 2 188
BC028526.1 - 2 174 3371464
BY347589.1 - 3 209

View Full Product Details