Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026932.4 | NP_081208.1 | 8, 9 | 1006 | |
GenBank mRNA | AK007491.1 | - | 8, 9 | 976 | |
AK134732.1 | - | 8, 9 | 961 | ||
AK167070.1 | - | 8, 9 | 961 | ||
AK168269.1 | - | 8, 9 | 933 | ||
BC054723.1 | - | 8, 9 | 989 | 5695176 | |
BC062876.1 | - | 8, 9 | 938 | 5686021 |