Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163476.1 | NP_001156948.1 | 3 | 332 | |
NM_027014.1 | NP_081290.1 | 3 | 332 | ||
GenBank mRNA | AK013116.1 | - | 3 | 325 | |
BC027537.1 | - | 3 | 309 | 3466154 | |
BQ553304.1 | - | 3 | 332 |