Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002427.3 | NP_002418.1 | 9 | 1273 | |
GenBank mRNA | AK222748.1 | - | 9 | 1273 | |
AK292211.1 | - | 9 | 1273 | ||
AK315341.1 | - | 9 | 1271 | ||
AY256443.1 | - | 9 | 1244 | ||
BC067522.1 | - | 9 | 1252 | 7002184 | |
BC067523.1 | - | 9 | 1252 | 7002185 | |
BC074807.2 | - | 9 | 1263 | 30915315 | |
BC074808.2 | - | 9 | 1263 | 30915529 | |
X75308.1 | - | 9 | 1248 | ||
X81334.1 | - | 9 | 1271 |