Gene Transcripts

Gene Symbol
Mier2
Entrez Gene ID
70427
Gene Name
mesoderm induction early response 1, family member 2
Gene Aliases
2700087H15Rik, mKIAA1193
Chromosome Location
Chr. 10: 79540245 - 79555631 on Build GRCm38
UniGene ID
Mm.334193
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027422.2 NP_081698.2 4 369
GenBank mRNA AK173128.1 - 3 525
BC067013.1 - 4 369 6810181

View Full Product Details