Gene Transcripts
- Gene Symbol
- Atad2
- Entrez Gene ID
- 70472
- Gene Name
- ATPase family, AAA domain containing 2
- Gene Aliases
- 2610509G12Rik
- Chromosome Location
- Chr. 15: 58094047 - 58135139 on Build GRCm38
- UniGene ID
- Mm.221758
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_027435.2 |
NP_081711.2
|
22 |
3188 |
|
GenBank mRNA |
AK029867.1 |
- |
15 |
2335 |
|
View Full Product Details