Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081368.1 | NP_001074837.1 | 4 | 792 | |
GenBank mRNA | AK017705.1 | - | 4 | 799 | |
AK039905.1 | - | 4 | 617 | ||
AK040787.1 | - | 4 | 764 | ||
AK043059.1 | - | 4 | 766 | ||
AK133804.1 | - | 4 | 733 | ||
BC029679.1 | - | 176 | |||
BC082558.1 | - | 4 | 761 | 30605020 |