Gene Transcripts

Gene Symbol
Ccdc93
Entrez Gene ID
70829
Gene Name
coiled-coil domain containing 93
Gene Aliases
4633402D15Rik, 9230102M16Rik
Chromosome Location
Chr. 1: 121431067 - 121506460 on Build GRCm38
UniGene ID
Mm.25788
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025156.2 NP_001020327.1 3 367
NM_029955.3 NP_084231.2 3 367
GenBank mRNA AK014621.1 - 3 341
AK020306.1 - 3 327
AK052300.1 - 3 332
AK082581.1 - 3 360
AK085701.1 - 3 326
AK142302.1 - 3 385
AK159415.1 - 3 335
BC054083.1 - 3 270 6467830
BY730410.1 - 3 385

View Full Product Details