Gene Transcripts

Gene Symbol
MSI1
Entrez Gene ID
4440
Gene Name
musashi RNA binding protein 1
Gene Aliases
Chromosome Location
Chr. 12: 120339655 - 120369180 on Build GRCh38
UniGene ID
Hs.158311
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002442.3 NP_002433.1 7, 8 549
XM_006719403.3 XP_006719466.1 7, 8 543
XM_006719404.3 XP_006719467.1 2, 3 328
XM_011538361.2 XP_011536663.1 7, 8 543
XM_011538362.2 XP_011536664.1 7, 8 543
XM_011538363.2 XP_011536665.1 7, 8 543
XM_011538364.2 XP_011536666.1 3, 4 1093
XM_011538365.2 XP_011536667.1 4, 5 212
XM_011538366.2 XP_011536668.1 4, 5 1220
XM_011538368.2 XP_011536670.1 2, 3 168
XM_011538370.2 XP_011536672.1 2, 3 94
XM_011538371.2 XP_011536673.1 2, 3 120
GenBank mRNA AB012851.1 - 7, 8 524
AB072590.1 - 4, 5 271
AB072591.1 - 4, 5 271
AB072592.1 - 4, 5 271
AB073212.1 - 4, 5 264
BX393884.2 - 7, 8 549

View Full Product Details