Gene Transcripts

Gene Symbol
Plbd2
Entrez Gene ID
71772
Gene Name
phospholipase B domain containing 2
Gene Aliases
1300012G16Rik, AU019810, P76
Chromosome Location
Chr. 5: 120483893 - 120503623 on Build GRCm38
UniGene ID
Mm.100065
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023625.4 NP_076114.2 10 1397
GenBank mRNA AF217003.1 - 10 1378
AK004973.1 - 10 1400
AK029813.1 - 10 1396
AK030234.1 - 10 1349
AK032930.1 - 10 1393
AK042089.1 - 1393
AK043608.1 - 10 1387
AK150201.1 - 10 1386
AK164220.1 - 10 1378
AK170387.1 - 10 1389
AK170631.1 - 10 1377
AK188200.1 - 2 137
AK209064.1 - 2 137
BC026395.1 - 10 1375
BC038605.1 - 10 1378 4921245
BC048826.1 - 10 1384 4196604
BC060095.1 - 6 3787

View Full Product Details