Gene Transcripts

Gene Symbol
Kif23
Entrez Gene ID
71819
Gene Name
kinesin family member 23
Gene Aliases
3110001D19Rik, C87313, CHO1, Knsl5, MKLP-1, MKLP1
Chromosome Location
Chr. 9: 61917277 - 61946812 on Build GRCm38
UniGene ID
Mm.259374
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024245.4 NP_077207.3 5 582
GenBank mRNA AK013939.1 - 5 520
AK030771.1 - 5 559
BC003363.1 - 5 555
BC047273.1 - 5 540 6440797

View Full Product Details