Gene Transcripts

Gene Symbol
Reep4
Entrez Gene ID
72549
Gene Name
receptor accessory protein 4
Gene Aliases
2700029E10Rik
Chromosome Location
Chr. 14: 70545218 - 70548935 on Build GRCm38
UniGene ID
Mm.46742
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_180588.2 NP_850919.1 7 1056
GenBank mRNA AK012301.1 - 7 953
AY562232.1 - 7 600
BC033929.1 - 7 684 1363539

View Full Product Details