Gene Transcripts

Gene Symbol
Tdrkh
Entrez Gene ID
72634
Gene Name
tudor and KH domain containing protein
Gene Aliases
2700091C21Rik, Tdrd2
Chromosome Location
Chr. 3: 94413239 - 94432975 on Build GRCm38
UniGene ID
Mm.331198
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028307.1 NP_082583.1 11 1533
GenBank mRNA BC049363.1 - 11 1533
BC057030.1 - 11 1549 6833743

View Full Product Details