Gene Transcripts

Gene Symbol
MYBL2
Entrez Gene ID
4605
Gene Name
MYB proto-oncogene like 2
Gene Aliases
B-MYB, BMYB
Chromosome Location
Chr. 20: 43667019 - 43716496 on Build GRCh38
UniGene ID
Hs.179718
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278610.1 NP_001265539.1 6 947
NM_002466.3 NP_002457.1 7 1019
GenBank mRNA AK223482.1 - 7 754
AK303249.1 - 6 852
AK314791.1 - 7 924
BC007585.1 - 7 944 3162656
BC053555.1 - 7 913 6140320
X13293.1 - 7 881

View Full Product Details