Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028419.2 | NP_082695.1 | 1 | 244 | |
GenBank mRNA | AK009158.1 | - | 1 | 249 | |
AK013761.1 | - | 1 | 245 | ||
AK050883.1 | - | 244 | |||
BC050937.1 | - | 1 | 264 | ||
BC058371.1 | - | 1 | 264 | 6819353 | |
DQ083330.1 | - | 1 | 233 |