Gene Transcripts

Gene Symbol
Rwdd3
Entrez Gene ID
66568
Gene Name
RWD domain containing 3
Gene Aliases
2510027J23Rik, 3110037C01Rik, AI035684, AI662458, Rsume, X2CR1
Chromosome Location
Chr. 3: 121155400 - 121171780 on Build GRCm38
UniGene ID
Mm.263163
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025637.3 NP_079913.2 5 839
NM_028456.3 NP_082732.2 3 623
GenBank mRNA AF439556.1 - 3 622
AF439557.1 - 4 829
AK014130.1 - 3 561
AK134288.1 - 3 614
BC037515.1 - 1 222
BC128287.1 - 3 622 40102432
BC128288.1 - 5 838
BC139340.1 - 3 622 8862362
BC139342.1 - 3 622 8862364

View Full Product Details