Gene Transcripts

Gene Symbol
Wdr75
Entrez Gene ID
73674
Gene Name
WD repeat domain 75
Gene Aliases
1300003A18Rik, 2410118I19Rik, C77608
Chromosome Location
Chr. 1: 45795153 - 45823619 on Build GRCm38
UniGene ID
Mm.270376
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028599.2 NP_082875.1 11, 12 1127
GenBank mRNA AK010772.1 - 11, 12 1127
AK152412.1 - 11, 12 1128
AK163221.1 - 11, 12 1101
AK168032.1 - 11, 12 1121
AK168163.1 - 11, 12 1129
AK169932.1 - 11, 12 1125
BC085177.1 - 11, 12 1157
BC117880.2 - 11, 12 1157 40096010
BC117881.1 - 11, 12 1154 40096014

View Full Product Details