Gene Transcripts

Gene Symbol
Spatc1
Entrez Gene ID
74281
Gene Name
spermatogenesis and centriole associated 1
Gene Aliases
1700084J23Rik, speriolin
Chromosome Location
Chr. 15: 76267835 - 76292577 on Build GRCm38
UniGene ID
Mm.159156
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028852.1 NP_083128.1 1 177
GenBank mRNA AB032199.1 - 1 177
AK006994.1 - 1 178
BC116725.1 - 1 144
BC118509.1 - 1 137 40106461

View Full Product Details