Gene Transcripts

Gene Symbol
Gvin1
Entrez Gene ID
74558
Gene Name
GTPase, very large interferon inducible 1
Gene Aliases
9130002C22Rik, 9830104F22Rik, Iigs1, VLIG, VLIG-1
Chromosome Location
Chr. 7: 106156535 - 106215377 on Build GRCm38
UniGene ID
Mm.458429
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039160.2 NP_001034249.2 2 7133
NM_029000.3 NP_083276.2 4 7327

Gene Symbol
Gm4070
Entrez Gene ID
100042856
Gene Name
predicted gene 4070
Gene Aliases
VLIG-1
Chromosome Location
Chr. 7: 105895118 - 105954002 on Build GRCm38
UniGene ID
Mm.425473
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243039.1 NP_001229968.1 4 7324
NM_001243040.1 NP_001229969.1 2 7130
GenBank mRNA AK046674.1 - 1 3220
AY167972.1 - 4 7313
AY167973.1 - 2 7103
AY167974.1 - 4 7297

View Full Product Details