Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029103.3 | NP_083379.2 | 4 | 514 | |
GenBank mRNA | AK014338.1 | - | 4 | 492 | |
AK131997.1 | - | 4 | 515 | ||
AK165646.1 | - | 4 | 490 | ||
AK167580.1 | - | 4 | 514 | ||
BC022761.1 | - | 4 | 520 | ||
BC038901.1 | - | 4 | 502 | ||
BC048616.1 | - | 3 | 577 | 6742574 |