Gene Transcripts
- Gene Symbol
- Mrpl32
- Entrez Gene ID
- 75398
- Gene Name
- mitochondrial ribosomal protein L32
- Gene Aliases
- 0610033O15Rik
- Chromosome Location
- Chr. 13: 14610301 - 14613037 on Build GRCm38
- UniGene ID
- Mm.36511
- Species
- Mus musculus
- Gene Symbol
- Gm6578
- Entrez Gene ID
- 625347
- Gene Name
- mitochondrial ribosomal protein L32 pseudogene
- Gene Aliases
- EG625347
- Chromosome Location
- Chr. 6: 12099518 - 12109580 on Build GRCm38
- UniGene ID
- Mm.389084
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC099426.1 |
- |
4 |
630 |
|
View Full Product Details