Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023815.4 | NP_076304.2 | 2 | 1076 | |
GenBank mRNA | AK028411.1 | - | 2 | 1077 | |
AK046261.1 | - | 2 | 1054 | ||
AK049291.1 | - | 2 | 1061 | ||
AK160279.1 | - | 2 | 1056 | ||
BC017155.1 | - | 2 | 1094 | 4910610 |