Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199954.1 | NP_001186883.1 | 6 | 1575 | |
NM_001614.3 | NP_001605.1 | 6 | 1456 | ||
NR_037688.1 | 6 | 1456 | |||
GenBank mRNA | AK291937.1 | - | 6 | 1389 | |
AK301175.1 | - | 7 | 1263 | ||
AK304637.1 | - | 7 | 1329 | ||
BC000292.2 | - | 6 | 1382 | 2819345 | |
BC001920.1 | - | 6 | 1345 | 2820489 | |
BC004223.2 | - | 5 | 1294 | ||
BC007442.2 | - | 6 | 1376 | 2962953 | |
BC009544.1 | - | 3 | 670 | ||
BC009848.2 | - | 6 | 1373 | 3939552 | |
BC010999.2 | - | 6 | 1375 | 3940861 | |
BC012050.1 | - | 6 | 1376 | 4554944 | |
BC015005.1 | - | 6 | 1371 | 4855463 | |
BC015695.2 | - | 6 | 1378 | 4842665 | |
BC015779.1 | - | 6 | 1352 | 4856294 | |
BC017450.1 | - | 5 | 1283 | ||
BC018774.1 | - | 6 | 1376 | 4869628 | |
BC018861.2 | - | 3 | 816 | ||
BC023548.2 | - | 4 | 983 | ||
BC053572.1 | - | 6 | 1369 | 3865767 | |
BC063495.1 | - | 5 | 1308 | ||
EU668331.1 | - | 3 | 888 | ||
M16247.1 | - | 3 | 888 | ||
X04098.1 | - | 6 | 1390 |