Gene Transcripts

Gene Symbol
Spata31
Entrez Gene ID
78124
Gene Name
spermatogenesis associated 31
Gene Aliases
4930458L03Rik, Aep1, Fam75a, Spata31a, Vad1.3
Chromosome Location
Chr. 13: 64917406 - 64923195 on Build GRCm38
UniGene ID
Mm.342895
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030047.2 NP_084323.2 3 413
GenBank mRNA AK019634.1 - 3 359
AK133249.1 - 3 414
AY842249.1 - 3 354

View Full Product Details