Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005385.3 | NP_005376.2 | 16 | 4361 | |
XM_005265173.1 | XP_005265230.1 | 16 | 4359 | ||
XM_006713171.1 | XP_006713234.1 | 17 | 4278 | ||
XM_006713173.2 | XP_006713236.1 | 17 | 4381 | ||
XM_011533747.2 | XP_011532049.1 | 17 | 7693 | ||
XM_011533750.2 | XP_011532052.1 | 18 | 4375 | ||
XM_017006474.1 | XP_016861963.1 | 17 | 4275 | ||
XM_017006475.1 | XP_016861964.1 | 17 | 4363 | ||
XM_017006476.1 | XP_016861965.1 | 11 | 8906 | ||
XM_017006477.1 | XP_016861966.1 | 11 | 5560 | ||
XM_017006478.1 | XP_016861967.1 | 11 | 5537 | ||
XM_017006479.1 | XP_016861968.1 | 10 | 6293 | ||
XM_017006480.1 | XP_016861969.1 | 18 | 4368 | ||
GenBank mRNA | BC144518.1 | - | 12 | 2816 | |
BX957215.1 | - | 8 | 5537 | ||
BX957218.1 | - | 18 | 4389 | ||
CR749415.1 | - | 8 | 5537 | ||
CR749571.1 | - | 18 | 4389 | ||
L04288.2 | - | 16 | 4402 |