Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030233.1 | NP_084509.1 | 10 | 1100 | |
GenBank mRNA | AB489137.1 | - | 10 | 1081 | |
AK019878.1 | - | 10 | 1101 | ||
AK030833.1 | - | 10 | 1106 | ||
AK031128.1 | - | 8 | 2948 | ||
AK033202.1 | - | 10 | 1111 | ||
AK035874.1 | - | 9 | 3144 | ||
AK132193.1 | - | 10 | 1101 | ||
AK134418.1 | - | 10 | 1108 | ||
BC019985.1 | - | 11 | 1450 | ||
BC028473.1 | - | 5 | 514 | ||
BC068127.1 | - | 10 | 1097 | ||
BC087944.1 | - | 10 | 1112 | 30314036 |