Gene Transcripts

Gene Symbol
Cttnbp2nl
Entrez Gene ID
80281
Gene Name
CTTNBP2 N-terminal like
Gene Aliases
AA552995, AA589392, AU018624, BC003236, mKIAA1433
Chromosome Location
Chr. 3: 105001915 - 105053146 on Build GRCm38
UniGene ID
Mm.200327
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163332.1 NP_001156804.1 4 302
NM_001163333.1 NP_001156805.1 4 294
NM_030249.4 NP_084525.1 4 410
GenBank mRNA AK029901.1 - 4 411
AK030438.1 - 4 275
AK051259.1 - 4 274
AK129359.1 - 4 300
AK145085.1 - 4 273
BC003236.1 - 4 264 3498078
BY195820.1 - 4 307
CV562515.1 - 4 294
EF410090.1 - 271

View Full Product Details