Gene Transcripts

Gene Symbol
Slmap
Entrez Gene ID
83997
Gene Name
sarcolemma associated protein
Gene Aliases
D330001L02Rik, Slap, mKIAA1601
Chromosome Location
Chr. 14: 26413168 - 26534624 on Build GRCm38
UniGene ID
Mm.36769
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032008.3 NP_114397.3 16 1705
GenBank mRNA AF304451.1 - 4 253
AK129403.1 - 16 1705
AK141597.1 - 17 2925
AK146685.1 - 4 219
BC021457.1 - 6 468 5064534

View Full Product Details